Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ASGQEFWPG QSAADILSGA ASRRRYLLYD VNPPEGFNLR RDVYIRIASL LKTLLKTEEW VLVLPPWGRL YHWQSPDIHQ VRIPWSEFFD LPSLNKNIPV IEYEQFIAES GGPFIDQVYV LQSYAEGWKE GTWEEKVDER PCIDQLLYSQ DKHEYYRGWF WGYEETRGLN VSCLSVQGSA SIVAPLLLRN TSARSVMLDR AENLLHDHYG GKEYWDTRRS MVFARHLREV GDEFRSRHLN STDDADRIPF QEDWMKMKVK LGSALGGPYL GVHLRRKDFI WGHRQDVPSL EGAVRKIRSL MKTHRLDKVF VATDAVRKEY EELKKLLPEM VRFEPTWEEL ELYKDGGVAI IDQWICAHAR FFIGTSVSTF SFRIHEEREI LGLDPKTTYN RFCGDQEKAC EQPTHWKITY (21-429 aa encoded by BC064623)
Fucose is typically found as a terminal modification of branched chain glycoconjugates, but it also exists in direct O-linkage to serine or threonine residues within cystine knot motifs in epidermal growth factor-like repeats or thrombospondin type-1 repeats. POFUT2 is an O-fucosyltransferase that use THBS type-1 repeats as substrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDP-fucose protein O-fucosyltransferase 2; O-FucT-2; Peptide-O-fucosyltransferase 2
Gene Aliases: 2310011G23Rik; AI256847; BC003494; C21orf80; FUT13; KIAA0958; POFUT2
UniProt ID: (Human) Q9Y2G5, (Mouse) Q8VHI3
Entrez Gene ID: (Human) 23275, (Rat) 309686, (Mouse) 80294
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support