Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MESTAYPLNL SLKEEEEEEE IQSRELEDGP ADMQKVRICS EGGWVPALFD EVAIYFSDEE WEVLTEQQKA LYREVMRMNY ETVLSLEFPF PKPDMITRLE GEEESQNSDE WQLQGGTSAE NEESDVKPPD WPNPMNATSQ FPQPQHFDSF GLRLPRDITE LPEWSEGYPF YMAMGFPGYD LSADDIAGKF QFSRGMRRSY DAGFKLMVVE YAESTNNCQA AKQFGVLEKN VRDWRKVKPQ LQNAHAMRRA FRGPKNGRFA LVDQRVAEYV RYMQAKGDPI TREAMQLKAL EIAQEMNIPE KGFKASLGWC RRMMRRYDLS LRHKVPVPQH LPEDLTEKLV TYQRSVLALR RAHDYEVAQM GNADETPICL EVPSRVTVDN QGEKPVLVKT PGREKLKITA MLGVLADGRK LPPYIILRGT YIPPGKFPSG MEIRCHRYGW MTEDLMQDWL EVVWRRRTGA VPKQRGMLIL NGFRGHATDS VKNSMESMNT DMVIIPGGLT SQLQVLDVVV YKPLNDSVRA QYSNWLLAGN LALSPTGNAK KPPLGLFLEW VMVAWNSISS ESIVQGFKKC HISSNLEEED DVLWEIESEL PGGGEPPKDC DTESMAESN
This gene encodes a 134 kDa protein named strumpellin that is predicted to have multiple transmembrane domains and a spectrin-repeat-containing domain. This ubiquitously expressed gene has its highest expression in skeletal muscle. The protein is named for Strumpell disease; a form of hereditary spastic paraplegia (HSP). Spastic paraplegias are a diverse group of disorders in which the autosomal dominant forms are characterized by progressive, lower extremity spasticity caused by axonal degeneration in the terminal portions of the longest descending and ascending corticospinal tracts. More than 30 loci (SPG1-33) have been implicated in hereditary spastic paraplegia diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KRAB box domain containing 2; Pogo transposable element with KRAB domain
Gene Aliases: BASS2; KIAA1513; KRBOX2; LST003; POGK; SLTP003
UniProt ID: (Human) Q9P215
Entrez Gene ID: (Human) 57645
Molecular Function:
viral or transposable element protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support