Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
POLE (Polepsi) is present in proliferating cells and is implicated in repair of UV-damaged DNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-5' exodeoxyribonuclease; DNA polymerase epsilon catalytic subunit A; DNA polymerase epsilon catalytic subunit protein; DNA polymerase II subunit A; polymerase (DNA directed), epsilon, catalytic subunit; polymerase (DNA) epsilon, catalytic subunit
Gene Aliases: CRCS12; FILS; POLE; POLE1
UniProt ID: (Human) Q07864
Entrez Gene ID: (Human) 5426
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support