Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAGNKGRGR AAYTFNIEAV GFSKGEKLPD VVLKPPPLFP DTDYKPVPLK TGEGEEYMLA LKQELRETMK RMPYFIETPE ERQDIERYSK RYMKVYKEEW IPDWRRLPRE MMPRNKCKKA GPKPKKAKDA GKGTPLTNTE DVLKKMEELE KRGDGEKSDE ENEEKEGSKE KSKEGDDDDD DDAAEQEEYD EEEQEEENDY INSYFEDGDD FGADSDDNMD EATY (1-223 aa encoded by BC141649)
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. POLR3G is a specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. It may direct with other members of the RPC3/POLR3C-RPC6/POLR3F-RPC7/POLR3G subcomplex RNA Pol III binding to the TFIIIB-DNA complex via the interactions between TFIIIB and POLR3F. It may be involved either in the recruitment and stabilization of the subcomplex within RNA polymerase III, or in stimulating catalytic functions of other subunits during initiation. POLR3G plays a key role in sensing and limiting infection by intracellular bacteria and DNA viruses. POLR3G acts as nuclear and cytosolic DNA sensor involved in innate immune response. POLR3G can sense non-self dsDNA that serves as template for transcription into dsRNA. The non-self RNA polymerase III transcripts, such as Epstein-Barr virus-encoded RNAs (EBERs) induce type I interferon and NF- Kappa-B through the RIG-I pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-directed RNA polymerase III 32 kDa polypeptide; DNA-directed RNA polymerase III subunit G; DNA-directed RNA polymerase III subunit RPC7; polymerase (RNA) III (DNA directed) polypeptide G (32kD); polymerase (RNA) III subunit G; RNA polymerase III 32 kDa apha subunit; RNA polymerase III 32 kDa subunit; RNA polymerase III subunit C7; RPC32; RPC32-alpha
Gene Aliases: POLR3G; RPC32; RPC7
UniProt ID: (Human) O15318
Entrez Gene ID: (Human) 10622
Molecular Function: DNA-directed RNA polymerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support