Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: DQIFLDGQEN KRSCLVDGLT DASSAFKVPR PGPDTLQFTV DVFHFANDSR NM*
This gene appears to have resulted from a fusion of DNA sequences derived from 2 distinct loci, specifically through the duplication of two internal exons from the POM121 gene and four 3' exons from the ZP3 gene. The 5' end of this gene is similar to the 5` coding region of the POM121 gene which encodes an integral nuclear pore membrane protein. However, the protein encoded by this gene lacks the nuclear pore localization motif. The 3' end of this gene is similar to the last 4 exons of the zona pellucida glycoprotein 3 (ZP3) gene and the encoded protein retains one zona pellucida domain. Multiple protein isoforms are encoded by transcript variants of this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: POM (POM121 homolog, rat) and ZP3 fusion; POM (POM121 rat homolog) and ZP3 fusion; POM-ZP3; POM-ZP3 fusion protein; POM121 and ZP3 fusion protein; POM121/ZP3 fusion protein
Gene Aliases: POM-ZP3; POM121; POMZP3
UniProt ID: (Human) Q6PJE2
Entrez Gene ID: (Human) 22932
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support