Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Glycosylated PON1 migrates at 42-45 kDa. 55 kDa is a dimmer of isoform CRA-b. The antibody has cross-reaction to PON2 and PON3.
Immunogen sequence: MAKLIALTL LGMGLALFRN HQSSYQTRLN ALREVQPVEL PNCNLVKGIE TGSEDLEILP NGLAFISSGL KYPGIKSFNP NSPGKILLMD LNEEDPTVLE LGITGSKFDV SSFNPHGIST FTDEDNAMYL LVVNHPDAKS TVELFKFQEE EKSLLHLKTI RHKLLPNLND IVAVGPEHFY GTNDHYFLDP YLQSWEMYLG LAWSYVVYYS PSEVRVVAEG FDFANGINIS PDGKYVYIAE LLAHKIHVYE KHANWTLTPL KSLDFNTLVD NISVDPETGD LWVGCHPNGM KIFFYDSENP PASEVLRIQN ILTEEPKVTQ VYAENGTVLQ GSTVASVYKG KLLIGTVFHK ALYCEL (1-355 aa encoded by B C074719)
The paraoxonase (PON) gene-family in humans has three members, PON1, PON2, and PON3, aligned next to each other on the long arm of chromosome 7q21.3-22.1. PON1 of these is synthesized primarily in the liver and a portion is secreted into the plasma, where it is associated with high density lipoproteins (HDL). And It hydrolyzes the active metabolites of several other organophosphorus insecticides (e.g., chlorpyrifos oxon, diazoxon), as well as nerve agents such as sarin, soman and VX (lethal nerve agent). The PON1 polymorphism may be responsible for neurodegeneration and considered to be an independent risk factor for Parkinson's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A esterase 1; A-esterase 1; Aromatic esterase 1; arylesterase 1; arylesterase B-type; EC 3.1.1.2; EC 3.1.1.81; EC 3.1.8.1; ESAesterase A; esterase A; K-45; paraoxonase B-type; PON 1; serum aryldiakylphosphatase; Serum aryldialkylphosphatase 1; Serum paraoxonase/arylesterase 1
Gene Aliases: ESA; MVCD5; PON; PON1
UniProt ID: (Human) P27169, (Mouse) P52430, (Rat) P55159
Entrez Gene ID: (Human) 5444, (Mouse) 18979, (Rat) 84024
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support