Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKSVIYHAL SQKEANDSDV QPSGAQRAEA FVRAFLKRST PRMSPQARED QLQRKAVVLE YFTRHKRKEK KKKAKGLSAR QRRELRLFDI KPEQQRYSLF LPLHELWKQY IRDLCSGLKP DTQPQMIQAK LLKADLHGAI ISVTKSKCPS YVGITGILLQ ETKHIFKIIT KEDRLKVIPK LNCVFTVETD GFISYIYGSK FQLRSSERSA KKFKAKGTID L (1-220 aa encoded by BC004438)
POP4 is a part of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends. It may function with RPP38 to coordinate the nucleolar targeting and/or assembly of RNase P.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPOP4; Ribonuclease P protein subunit p29
Gene Aliases: POP4; RPP29
UniProt ID: (Human) O95707
Entrez Gene ID: (Human) 10775
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support