Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ILTDHQYLE RTPLCAILKQ KAPQQYRIRA KLRSYKPRRL FQSVKLHCPK CHLLQEVPHE GDLDIIFQDG ATKTPDVKLQ NTSLYDSKIW TTKNQKGRKV AVHFVKNNGI LPLSNECLLL IEGGTLSEIC KLSNKFNSVI PVRSGHEDLE LLDLSAPFLI QGTIHHYGCK QCSSLRSIQN LNSLVDKTSW IPSSVAEALG IVPLQYVFVM TFTLDDGTGV LEAYLMDSDK FFQIPASEVL MDDDLQKSVD MIMDMFCPPG IKIDAYPWLE CFIKSYNVTN GTDNQICYQI FDTTVAEDVI (336-634 aa encoded by BC002923)
POT1 is a component of the telomerase ribonucleoprotein (RNP) complex that is essential for the replication of chromosome termini. It is a component of the double-stranded telomeric DNA-binding TRF1 complex which is involved in the regulation of telomere length by cis-inhibition of telomerase. Pot1 also acts as a single-stranded telomeric DNA-binding protein and thus may act as a downstream effector of the TRF1 complex and may transduce information about telomere maintenance and/or length to the telomere terminus. It binds to two or more telomeric single-stranded 5'-TTAGGG-3' repeats (G-strand) and with high specificity to a minimal telomeric single-stranded 5'-TAGGGTTAG-3' sequence. Its activity is TERT dependent but it does not increase TERT activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp586D211; hPot1; mPot1; POT 1; POT-1; POT1-like telomere end-binding protein; protection of telomeres 1 homolog; Protection of telomeres protein 1
Gene Aliases: 1500031H18Rik; AI851169; CMM10; GLM9; HPOT1; POT1; Pot1a
UniProt ID: (Human) Q9NUX5, (Mouse) Q91WC1
Entrez Gene ID: (Human) 25913, (Mouse) 101185
Molecular Function: DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support