Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MMAMNSKQPF GMHPVLQEPK FSSLHSGSEA MRRVCLPAPQ LQGNIFGSFD ESLLARAEAL AAVDIVSHGK NHPFKPDATY HTMSSVPCTS TSSTVPISHP AALTSHPHHA VHQGLEGDLL EHISPTLSVS GLGAPEHSVM PAQIHPHHLG AMGHLHQAMG MSHPHTVAPH SAMPACLSDV ESDPRELEAF AERFKQRRIK LGVTQADVGA ALANLKIPGV GSLSQSTICR FESLTLSHNN MIALKPVLQA WLEEAEAAYR EKNSKPELFN GSERKRKRTS IAAPEKRSLE AYFAIQPRPS SEKIAAIAEK LDLKKNVVRV WFCNQRQKQK RMKYSAVH
POU4F3 encodes a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. This protein is found in the retina and may play a role in determining or maintaining the identities of a small subset of visual system neurons. Defects in this gene are the cause of non-syndromic sensorineural deafness autosomal dominant type 15.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-3C; Brain-specific homeobox/POU domain protein 3C; brn-3C; MGC138412; POU domain, class 4, transcription factor 3
Gene Aliases: BRN3C; DFNA15; POU4F3
UniProt ID: (Human) Q15319
Entrez Gene ID: (Human) 5459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support