Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPLRVDTLTW LSTQAAPGRV MVWPAVRPGI CPGPDVWRIP LGPLPHEFRG WIAPCRPRLG ASEAGDWLRR PSEGALPGPY IALRSIPKLP PPEDISGILK ELQQLAKELR QKRLSLGYSQ ADVGIAVGAL FGKVLSQTTI CRFEAQQLSV ANMWKLRPLL KKWLKEVEAE NLLGLCKMEM ILQQSGKWRR ASRERRIGNS LEKFFQRCPK PTPQQISHIA GCLQLQKDVV RVWFYNRSKM GSRPTNDASP REIVGTAGPP CPGAPVCFHL GLGLPVDIPH YTRLYSAGVA HSSAPATTLG LLRF
Transcription factor that binds preferentially to the octamer motif (5'-ATGTTAAT-3'). May exert a regulatory function in meiotic events that are required for terminal differentiation of male germ cell.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: POU domain, class 5, transcription factor 2; Sperm 1 POU domain transcription factor; SPRM-1
Gene Aliases: POU5F2; SPRM-1; SPRM1
UniProt ID: (Human) Q8N7G0
Entrez Gene ID: (Human) 134187
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support