Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a HEAT-like repeat-containing protein. The HEAT repeat is a tandemly repeated, 37-47 amino acid long module occurring in a number of cytoplasmic proteins. Arrays of HEAT repeats form a rod-like helical structure and appear to function as protein-protein interaction surfaces. The repeat-containing region of this protein has some similarity to the constant regulatory domain of the protein phosphatase 2A PR65/A subunit. The function of this particular gene product has not been determined. Alternative splicing has been observed for this gene and two transcript variants encoding distinct isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cilia and flagella associated protein 14; HEAT-like repeat-containing protein; protein phosphatase 4, regulatory subunit 4; Serine/threonine-protein phosphatase 4 regulatory subunit 4
Gene Aliases: 8430415E04Rik; CFAP14; KIAA1622; mKIAA1622; PP4R4; PPP4R4; RGD1560932
UniProt ID: (Human) Q6NUP7, (Mouse) E9QKU6
Entrez Gene ID: (Human) 57718, (Mouse) 74521, (Rat) 500711
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support