Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Hydrolyzes inorganic pyrophosphate. This activity is essential for correct regulation of mitochondrial membrane potential, and mitochondrial organization and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20459; Inorganic pyrophosphatase 2, mitochondrial; PPase 2; Pyrophosphatase SID6-306; Pyrophosphate phospho-hydrolase 2
Gene Aliases: HSPC124; PPA2; SID6-306
UniProt ID: (Human) Q9H2U2
Entrez Gene ID: (Human) 27068
Molecular Function:
pyrophosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support