Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: KIIISKHDGG SPSIKAKVTL QKPGESSGEG LRPQMASSKN YGSPLISGST PKHERGSPSH SKSPAYTPQN LDSESESGSS IAEKSYQNSP SSDDGIRPLP
Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator-recruited cofactor 205 kDa component; ARC205; Mediator complex subunit 1; Mediator of RNA polymerase II transcription subunit 1; p53 regulatory protein RB18A; PBP; Peroxisome proliferator-activated receptor-binding protein; PPAR binding protein; PPAR-binding protein; PPARG binding protein; PPRB; Thyroid hormone receptor-associated protein complex 220 kDa component; thyroid hormone receptor-associated protein complex component TRAP220; thyroid receptor interacting protein 2; Thyroid receptor-interacting protein 2; TR-interacting protein 2; Trap220; TRIP-2; vitamin D receptor-interacting protein 230 kD; Vitamin D receptor-interacting protein complex component DRIP205
Gene Aliases: ARC205; CRSP1; CRSP200; DRIP205; DRIP230; MED1; PBP; PPARBP; PPARGBP; RB18A; TRAP220; TRIP2
UniProt ID: (Human) Q15648
Entrez Gene ID: (Human) 5469
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support