Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • PPARGC1A Antibodies

            Invitrogen

            PPARGC1A Polyclonal Antibody

            View all (2) PPARGC1A antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # PA5-144904
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            590.00
            100 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite PPARGC1A Polyclonal Antibody

            • Antibody Testing Data (1)
            Application
            PPARGC1A Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            PPARGC1A Antibody (PA5-144904) in ICC/IF

            Immunocytochemistry analysis of PPARGC1A in HeLa cells. Samples were incubated with PPARGC1A polyclonal antibody (Product # PA5-144904). Shows localization to nucleoplasm.
            View Product

            PPARGC1A Antibody in Immunocytochemistry (ICC/IF)

            Product Details

            PA5-144904

            Applications
            Tested Dilution
            Publications

            Immunocytochemistry (ICC/IF)

            0.25-2 µg/mL
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 300-381.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.4 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2, with 40% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_3092037

            Product Specific Information

            Immunogen sequence: PPHKANQDNP FRASPKLKSS CKTVVPPPSK KPRYSESSGT QGNNSTKKGP EQSELYAQLS KSSVLTGGHE ERKTKRPSLR LF

            Antigen sequence identity: Mouse 87%; Rat 88%

            Target Information

            PGC1 alpha is a transcriptional coactivator that regulates the genes involved in energy metabolism. It interacts with PPARgamma, which permits it's interaction with multiple transcription factors. PGC-1 Alpha can interact with, and regulate the activities of, cAMP response element binding protein (CREB) and nuclear respiratory factors (NRFs). It provides a direct link between external physiological stimuli and the regulation of mitochondrial biogenesis, and is a major factor that regulates muscle fiber type determination. It may be also involved in controlling blood pressure, regulating cellular cholesterol homoeostasis, and the development of obesity.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: L-PGC-1alpha; Ligand effect modulator 6; ligand effect modulator-6; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT; Peroxisome proliferator-activated receptor gamma coactivator 1-alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 alpha; pgc-1 alpha; PGC-1-alpha; PPAR gamma coactivator variant form; PPARgamma coactivator 1alpha; PPARGC-1-alpha

            View more View less

            Gene Aliases: LEM6; PGC-1(alpha); PGC-1alpha; PGC-1v; PGC1; PGC1A; PPARGC1; PPARGC1A

            View more View less

            UniProt ID: (Human) Q9UBK2

            View more View less

            Entrez Gene ID: (Human) 10891

            View more View less

            Function(s)

            nucleotide binding RNA polymerase II transcription cofactor activity DNA binding transcription coactivator activity RNA binding protein binding transcription factor binding ligand-dependent nuclear receptor binding estrogen receptor binding ligand-dependent nuclear receptor transcription coactivator activity chromatin DNA binding ubiquitin protein ligase binding peroxisome proliferator activated receptor binding alpha-tubulin binding sequence-specific DNA binding androgen receptor binding promoter-specific chromatin binding transcription cofactor

            Process(es)

            response to reactive oxygen species mitophagy temperature homeostasis cellular glucose homeostasis negative regulation of protein phosphorylation response to dietary excess response to ischemia galactose metabolic process gluconeogenesis regulation of transcription, DNA-templated transcription initiation from RNA polymerase II promoter mRNA processing protein complex assembly mitochondrion organization positive regulation of transcription from RNA polymerase II promoter by pheromones aging digestion circadian rhythm androgen metabolic process RNA splicing response to cold positive regulation of mitochondrion organization skeletal muscle atrophy response to muscle activity response to electrical stimulus involved in regulation of muscle adaptation negative regulation of smooth muscle cell migration fatty acid oxidation cerebellum development respiratory electron transport chain androgen receptor signaling pathway forebrain development circadian regulation of gene expression cellular response to oxidative stress positive regulation of histone acetylation cellular response to potassium ion response to statin response to drug response to starvation regulation of circadian rhythm response to leucine negative regulation of neuron apoptotic process cellular respiration positive regulation of gluconeogenesis negative regulation of glycolytic process positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of fatty acid oxidation positive regulation of smooth muscle cell proliferation negative regulation of smooth muscle cell proliferation protein stabilization brown fat cell differentiation positive regulation of sequence-specific DNA binding transcription factor activity flavone metabolic process adipose tissue development cellular response to lipopolysaccharide cellular response to nitrite cellular response to caffeine cellular response to fructose stimulus cellular response to glucose stimulus cellular response to interleukin-6 cellular response to tumor necrosis factor cellular response to follicle-stimulating hormone stimulus cellular response to estradiol stimulus cellular response to hypoxia cellular response to transforming growth factor beta stimulus response to epinephrine response to norepinephrine negative regulation of mitochondrial fission cellular response to thyroid hormone stimulus negative regulation of neuron death response to metformin positive regulation of cellular respiration positive regulation of mitochondrial DNA metabolic process positive regulation of muscle tissue development positive regulation of glomerular visceral epithelial cell apoptotic process cellular response to ionomycin cellular response to resveratrol response to methionine adaptive thermogenesis positive regulation of progesterone biosynthetic process negative regulation of receptor activity regulation of N-methyl-D-aspartate selective glutamate receptor activity positive regulation of energy homeostasis positive regulation of ATP biosynthetic process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-z7php:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0