Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MGPGPRLLLP LVLCVGLGAL VFSSGAEGFR KRGPSVTAKV FFDVRIGDKD VGRIVIGLFG KVVPKTVENF VALATGEKGY GYKGSKFHRV IKDFMIQGGD ITTGDGTGGV SIYGETFPDE NFKL
PPIC is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. Similar to other PPIases, this protein can bind immunosuppressant cyclosporin A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclophilin C; parvulin; Peptidyl-prolyl cis-trans isomerase C; peptidylprolyl isomerase C (cyclophilin C); PPIase C; Rotamase C
Gene Aliases: CYPC; PPIC
UniProt ID: (Human) P45877
Entrez Gene ID: (Human) 5480
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support