Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Human PPIL1.
This gene is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cyclophilin like 1; cyclophilin-related gene 1; peptidyl-prolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase-like 1; peptidylprolyl isomerase (cyclophilin)-like 1; PPIase; Rotamase PPIL1
Gene Aliases: CGI-124; CYPL1; hCyPX; PPIase; PPIL1; UNQ2425/PRO4984
UniProt ID: (Human) Q9Y3C6
Entrez Gene ID: (Human) 51645
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support