Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDNVLIDDR RIHVDFSQSV AKVKWKGKGG KYTKSDFKEY EKEQDKPPNL VLKDKVKPKQ DTKYDLILDE QAEDSKSSHS HTSKKHKKKT HHCSEEKEDE DYMPIKNTNQ DIYREMGFGH YEEEESCWEK QKSEKRDRTQ NRSRSRSRER DGHYSNSHKS KYQTDLYERE RSKKRDRSRS PKKSKDKEKS KYR (301-492 aa encoded by BC016984)
This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclophilin-like protein PPIL4; cyclophilin-type peptidyl-prolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase-like 4; peptidylprolyl isomerase (cyclophilin)-like 4; peptidylprolyl isomerase-like 4; PPIase; Rotamase PPIL4; serologically defined breast cancer antigen NY-BR-18
Gene Aliases: 3732410E19Rik; 3830425H19Rik; AI788954; AW146233; HDCME13P; PPIase; PPIL4
UniProt ID: (Human) Q8WUA2, (Mouse) Q9CXG3
Entrez Gene ID: (Human) 85313, (Mouse) 67418, (Rat) 361449
Molecular Function:
chaperone
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support