Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: EEEEDDDEEK APVTLLDAQS LAQSFFNRLW EVAGQWQKQV PLAARASQRQ WLVSIHAIRN TRRKMEDRHV SLPSFNQLFG LSDPVNRAYF AVFDGHGGVD AARYAAVHVH TNAARQPELP TDPEGALREA FRRTDQMFLR KAKRERLQSG TTGVCALIAG ATLHVAWLGD SQVILVQQGQ VVKLMEPHRP ERQDEKARIE ALGGFVSHMD CWRVNGTLAV SRAIGDVFQK PYVSGEADAA SRALTGSEDY LLLACDGFFD VVPHQEVVGL VQSHLTRQQG SGLRVAEELV AAARERGSHD NITVMVVFLR DPQELLEGGN QGEGDPQAEG RRQDLPSSLP EPETQAPPRS
PPM1F is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase can interact with Rho guanine nucleotide exchange factors (PIX), and thus block the effects of p21-activated kinase 1 (PAK), a protein kinase mediating biological effects downstream of Rho GTPases. Calcium/calmodulin-dependent protein kinase II gamma (CAMK2G/CAMK-II) is found to be one of the substrates of this phosphatase. The overexpression of this phosphatase or CAMK2G has been shown to mediate caspase-dependent apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ca(2+)/calmodulin-dependent protein kinase phosphatase; CaM-kinase phosphatase; hFem-2; Partner of PIX 2; PP2C phosphatase; Protein fem-2 homolog; Protein phosphatase 1F; protein phosphatase 1F (PP2C domain containing); protein phosphatase, Mg2+/Mn2+ dependent, 1F
Gene Aliases: CAMKP; CaMKPase; FEM-2; hFEM-2; KIAA0015; POPX2; PPM1F
UniProt ID: (Human) P49593
Entrez Gene ID: (Human) 9647
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support