Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Dephosphorylates CDKN1B at 'Thr-187', thus removing a signal for proteasomal degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neurite extension-related protein phosphatase related to PP2C; Protein phosphatase 1H; protein phosphatase 1H (PP2C domain containing); protein phosphatase, Mg2+/Mn2+ dependent, 1H; ras homolog gene family, member C like 1
Gene Aliases: A430075L18Rik; ARHCL1; C030002B11Rik; KIAA1157; NERPP-2C; PPM1H; URCC2
UniProt ID: (Human) Q9ULR3, (Mouse) Q3UYC0
Entrez Gene ID: (Human) 57460, (Mouse) 319468
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support