Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The antibody is specific to PPP1R11.
This gene encodes a specific inhibitor of protein phosphatase-1 (PP1) with a differential sensitivity toward the metal-independent and metal-dependent forms of PP1. The gene is located within the major histocompatibility complex class I region on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase PPP1R11; HCG V; Hemochromatosis candidate gene V protein; inhibitor-3; Protein phosphatase 1 regulatory subunit 11; protein phosphatase 1, regulatory (inhibitor) subunit 11; protein phosphatase 1, regulatory (inhibitor) subunit 11, isoform 1; Protein phosphatase inhibitor 3; T-complex testis expressed protein 5; t-complex testis-expressed 5; t-complex-associated-testis-expressed 5; Tctex-5
Gene Aliases: 1500041B02Rik; AV117883; CFAP255; HCG-V; HCGV; IPP3; PPP1R11; TCTE5; Tctex-5; TCTEX5
UniProt ID: (Human) O60927, (Mouse) Q8K1L5
Entrez Gene ID: (Human) 6992, (Mouse) 76497
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support