Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: ELRKRLNLEE WILEQLTRLY DCQEEEIPEL EIDVDELLDM ESDDARAARV KELLVDCYKP TEAFISGLLD KIRGMQ
This gene is thought to play a role in a complex developmental pathway and the regulation of genes involved in cerebellar development. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: phospholipase C beta-3 neighboring gene protein; Phospholipase C-beta-3 neighbouring gene protein; Protein phosphatase 1 regulatory subunit 14B; protein phosphatase 1, regulatory (inhibitor) subunit 14B
Gene Aliases: PHI-1; PLCB3N; PNG; PPP1R14B; SOM172
UniProt ID: (Human) Q96C90
Entrez Gene ID: (Human) 26472
Molecular Function:
phosphatase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support