Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: PRPLTSSVMP VDVAMRLCLA HSPPVKSFLG PYDEFQRRHF VNKLKPLKSC LNIKHKAKSQ NDWKCSHNQA KKRVVFADSK GLSLTAIHVF SDLPEEPAWD LQFDLLDL
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
Protein phosphatase-1 (PP1; see MIM 176875) participates in the regulation of a wide variety of cellular functions by reversible protein phosphorylation. The ability of PP1 to regulate diverse functions resides in its capacity to interact with a variety of regulatory subunits that may target PP1 to specific subcellular locations, modulate its substrate specificity, and allow its activity to be responsive to extracellular signals. Several targeting subunits of PP1 have been identified, including PPP1R5, the glycogen-binding subunits PPP1R3 (MIM 600917) and PPP1R4, and the nuclear inhibitor of PP1 (PPP1R8; MIM 602636).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Phosphatase 1, regulatory inhibitor subunit 5; PP1 subunit R5; PPP1R3C; Protein phosphatase 1 regulatory subunit 3C; Protein phosphatase 1 regulatory subunit 5; protein phosphatase 1, regulatory (inhibitor) subunit 3C; protein phosphatase 1, regulatory subunit 3C; Protein targeting to glycogen; PTG
Gene Aliases: PPP1R3C; PPP1R5
UniProt ID: (Human) B2R7X0
Entrez Gene ID: (Human) 5507
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support