Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
It is not recommended to aliquot this antibody.
This antibody is not cross-reactive with other PP2A subunits.
The PR65 subunit of protein phosphatase 2A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. Upon interaction with GNA12 promotes dephosphorylation of microtubule associated protein TAU/MAPT. Required for proper chromosome segregation and for centromeric localization of SGO1 in mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 65 kDa regulatory subunit of protein phosphatase; alpha isoform of regulatory subunit A, protein phosphatase 2; CG17291-PA; CG17291-PB; CG17291-PC; CG17291-PD; CG17291-PE; Medium tumor antigen-associated 61 kDa protein; MGC786; PP2A 65kD subunit A; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform R1-alpha; Pp2A-29B-PA; Pp2A-29B-PB; Pp2A-29B-PC; Pp2A-29B-PD; Pp2A-29B-PE; PP2Ac; PP2Calpha; PR65; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform; protein phosphatase 2 regulatory subunit A, alpha; protein phosphatase 2, regulatory subunit A, alpha; protein phosphatase 2A; protein phosphatase PP2A; Protein phosphatase PP2A regulatory subunit A; protein phosphate 2A; regulatory subunit A of type 2A protein phosphatase; serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alpha isoform; serine/threonine protein phosphatase A subunit type 2A; Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; Serine/threonine-protein phosphatase PP2A 65 kDa regulatory subunit; testicular secretory protein Li 1
Gene Aliases: 29B; 6330556D22Rik; A/PR65; CG13383; CG17291; CG33297; Dmel\CG17291; Dmel_CG17291; dPP2A A; DPR65; MRD36; PP2A; PP2A 29B; PP2A A; PP2A-29B; PP2A-A; PP2A-Aalpha; Pp2A29B; PP2A[A]; PP2AAALPHA; PPP2R1A; PR65; PR65A
UniProt ID: (Human) P30153, (Fruit fly) P36179, (Mouse) Q76MZ3
Entrez Gene ID: (Human) 5518, (Fruit fly) 2768940, (Mouse) 51792, (Rat) 117281
Molecular Function:
phosphatase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support