Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAAADGDDSL YPIAVLIDEL RNEDVQLRLN SIKKLSTIAL ALGVERTRSE LLPFLTDTIY DEDEVLLALA EQLGTFATLV GGPEYVHCLL PPLESLATVE ETVVRDKAVE SLRAISHEHS PSDLEAHFVP LVKRLAGGDW FTSRTSACGL FSVCYPRVSS AVKAELRQYF RNLCSDDTPM VRRAAASKLG EFAKVLELDN VKSEIIPMFS NLASDEQDSV RLLAVEACVN IAQLLPQEDL EALVMPTLRQ AAEDKSWRVR YMVADKFTEL QKAVGPEITK TDLVPAFQNL MKDCEAEVRA AASHKVKEFC ENLSADCREN VIMSQILPCI KELVSDANQH VKSALASVIM GLSPILGKDN TIEHLLPLFL AQLKDECPEV RLNIISNLDC VNEVIGIRQL SQSLLPAIVE LAEDAKWRVR LAIIEYMPLL AGQLGVEFFD EKLNSLCMAW LVDHVYAIRE AATSNLKKLV EKFGKEWAHA TIIPKVLAMS GDPNYLHRMT TLFCINVLSE VCGQDITTKH MLPTVLRMAG DPVANVRFNV AKSLQKIGPI LDNSTLQSEV KPILEKLTQD QDVDVKYFAQ EALTVLSLA
The PR65 subunit of protein phosphatase 2A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. Upon interaction with GNA12 promotes dephosphorylation of microtubule associated protein TAU/MAPT. Required for proper chromosome segregation and for centromeric localization of SGO1 in mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Medium tumor antigen-associated 61 kDa protein; MGC786; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform R1-alpha; PP2Ac; PP2Calpha; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 regulatory subunit A, alpha; protein phosphatase 2, regulatory subunit A, alpha; serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alpha isoform; Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; testicular secretory protein Li 1
Gene Aliases: MRD36; PP2A-Aalpha; PP2AAALPHA; PPP2R1A; PR65A
UniProt ID: (Human) P30153
Entrez Gene ID: (Human) 5518
Molecular Function:
phosphatase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support