Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDLDGDGALS MFELEYFYEE QCRRLDSMAI EALPFQDCLC QMLDLVKPRT EGKITLQDLK RCKLANVFFD TFFNIEKYLD HEQKEQISLL RDGDSGGPEL SDWEKYAAEE YDILVAEETA GEPWEDGFEA ELSPVEQKLS ALRSPLAQRP FFEAPSPLGA VDLYEYACGD EDLEPL
Protein phosphatase 2 is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B''/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B'' family. The B'' family has been further divided into subfamilies. The product of this gene belongs to the beta subfamily of regulatory subunit B''. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NY-REN-8; NY-REN-8 antigen; PP2A subunit B isoform PR48; PP2A, subunit B, PR48 isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B'', beta; protein phosphatase 2 regulatory subunit B'', beta; protein phosphatase 2, regulatory subunit B'', beta; Protein phosphatase 2A 48 kDa regulatory subunit; serine/threonine protein phosphatase 2A, 48kDa regulatory subunit B; Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit beta
Gene Aliases: NYREN8; PPP2R3B; PPP2R3L; PPP2R3LY; PR48
UniProt ID: (Human) Q9Y5P8
Entrez Gene ID: (Human) 28227
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support