Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Calmodulin-dependent protein phosphatase, calcineurin, is involved in a wide range of biologic activities, acting as a Ca(2+)-dependent modifier of phosphorylation status. In testis, the motility of the sperm is thought to be controlled by cAMP-dependent phosphorylation and a unique form of calcineurin appears to be associated with the flagellum. The calcineurin holoenzyme is composed of catalytic and regulatory subunits of 60 and 18 kD, respectively. At least 3 genes, calcineurin A-alpha, calcineurin A-beta, and calcineurin A-gamma (CALNA3), have been cloned for the catalytic subunit. These genes have been identified in humans, mice, and rats, and are highly conserved between species (90 to 95% amino acid identity).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcineurin A gamma; Calcineurin, testis-specific catalytic subunit; Calmodulin-dependent calcineurin A subunit gamma isoform; CAM-PRP catalytic subunit; protein phosphatase 2B, catalytic subunit, gamma isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, gamma isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, gamma isoform (calcineurin A gamma); protein phosphatase 3 catalytic subunit gamma; protein phosphatase 3, catalytic subunit, gamma isoform; protein phosphatase 3, catalytic subunit, gamma isozyme; Serine/threonine-protein phosphatase 2B catalytic subunit gamma isoform
Gene Aliases: CALNA3; Calnc; CNA3; PP2BA gamma; PP2Bgamma; PPP3CC
UniProt ID: (Human) P48454, (Mouse) P48455
Entrez Gene ID: (Human) 5533, (Mouse) 19057, (Rat) 171378
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support