Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: MAEISDLDRQ IEQLRRCELI KESEVKALCA KAREILVEES NVQRVDSPVT VCGDIHGQFY DLKELFRVGG DVPETNYLFM GDFVDRGFYS VETFLLLLAL KVRYPDRITL IRGNHESRQI TQVYGFYDEC LRKYGSVTVW RYCTEIFDYL SLSAIIDGKI FCVHGGLSPS IQTLDQIRTI DRKQEVPHDG PMCDLLWSDP EDTTGWGVSP RGAGYLFGSD VVAQFNAAND IDMICRAHQL VMEGYKWHFN ETVLTVWSAP NYCYRCGNVA AILELDEHLQ KDFIIFEAAP QETRGIPSKK PVADYFL; Positive Samples: 293T, NIH/3T3, MCF-7, HO-8910PM, Mouse testis, Mouse spleen; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center
The protein encoded by this gene is one subunit of a trimeric complex, forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes. Subunit A associates with a tight dimer composed of the B and C subunits, resulting in a trimer that binds to DNA with high specificity and affinity. The sequence specific interactions of the complex are made by the A subunit, suggesting a role as the regulatory subunit. In addition, there is evidence of post-transcriptional regulation in this gene product, either by protein degradation or control of translation. Further regulation is represented by alternative splicing in the glutamine-rich activation domain, with clear tissue-specific preferences for the two isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: P PPX; PP-X; pp4; PP4C; protein phosphatase 4 (formerly X), catalytic subunit; protein phosphatase 4, catalytic subunit; Protein phosphatase X; protein phosphatase X, catalytic subunit; Serine/threonine-protein phosphatase 4 catalytic subunit
Gene Aliases: 1110002D08Rik; AU016079; PP-X; PP4; PP4C; PPH3; PPP4; PPP4C; PPX
UniProt ID: (Human) P60510, (Rat) Q5BJ92, (Mouse) P97470
Entrez Gene ID: (Human) 5531, (Rat) 171366, (Mouse) 56420
Molecular Function: protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support