Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MNTSHLLALL LPKSPQGENR SKPLGTPYNF SEHCQDSVDV MVFIVTSYSI ETVVGVLGNL CLMCVTVRQK EKANVTNLLI ANLAFSDFLM CLLCQPLTAV YTIMDYWIFG ETLCKMSAFI QCMSVTVSIL SLVLVALERH QLIINPTGWK PSISQAYLGI VLIWVIACVL SLPFLANSIL ENVFHKNHSK ALEFLADKVV CTESWPLAHH RTIYTTFLLL FQYCLPLGFI LVCYARIYRR LQRQGRVFHK GTYSLRAGHM KQVNVVLVVM VVAFAMLWLP LHVFNSLEDW HHEAIPICHG NLIFLVCHLL AMASTCVNPF IYGFLNTNFK KEIKALVLTC QQSAPLEESE HLPLSTVHTE VSKGSLRLSG RSNPI
Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is PP, PP (2-36) and [Ile-31, Gln-34] PP > [Pro-34] PYY > PYY and [Leu-31, Pro-34] NPY > NPY > PYY (3-36) and NPY (2-36) > PP (13- 36) > PP (31-36) > NPY free acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuropeptide Y receptor type 4; NPY4-R; Pancreatic polypeptide receptor 1; PP1
Gene Aliases: NPY4-R; NPY4R; PP1; PPYR1; Y4
UniProt ID: (Human) P50391
Entrez Gene ID: (Human) 5540
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support