Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive test controls include: Mouse liver.
Immunogen sequence: MAHRPPSPAL ASVLLALLLS GAARAAEIVG GHEAQPHSRP YMASLQMRGN PGSHFCGGTL IHPSFVLTAA HCLRDIPQRL VNVVLGAHNV RTQEPTQQHF
Serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) (PubMed:3198760, PubMed:2033050, PubMed:28240246). By cleaving and activating receptor F2RL1/PAR-2, enhances endothelial cell barrier function and thus vascular integrity during neutrophil transendothelial migration (PubMed:23202369). May play a role in neutrophil transendothelial migration, probably when associated with CD177 (PubMed:22266279). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGP7; azurophil granule protein 7; C-ANCA antigen; Leukocyte proteinase 3; Myeloblastin; Neutrophil proteinase 4; NP-4; P29; PR-3; Proteinase 3; serine proteinase, neutrophil; Wegener autoantigen; Wegener granulomatosis autoantigen
Gene Aliases: ACPA; AGP7; C-ANCA; CANCA; MBN; MBT; mPR3; NP-4; NP4; P29; PR-3; PR3; PRTN3
UniProt ID: (Human) P24158, (Mouse) Q61096
Entrez Gene ID: (Human) 5657, (Mouse) 19152
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support