Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
In direct ELISAs, no cross-reactivity with recombinant mouse PRDM14, recombinant human (rh) ZNF24, rhZNF143, rhZNF206 or rhZNF281 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
PRDM14 encodes a member of the PRDI-BF1 and RIZ homology domain containing (PRDM) family of transcriptional regulators. The encoded protein may possess histone methyltransferase activity and plays a critical role in cell pluripotency by suppressing the expression of differentiation marker genes. Expression of this gene may play a role in breast cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PR domain containing 14; PR domain zinc finger protein 14; PR domain-containing 14; PR domain-containing protein 14
Gene Aliases: PFM11; PRDM14
UniProt ID: (Human) Q9GZV8
Entrez Gene ID: (Human) 63978
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support