Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Predicted Reactivity: Mouse (82%), Bovine (91%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PRDM14 encodes a member of the PRDI-BF1 and RIZ homology domain containing (PRDM) family of transcriptional regulators. The encoded protein may possess histone methyltransferase activity and plays a critical role in cell pluripotency by suppressing the expression of differentiation marker genes. Expression of this gene may play a role in breast cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PR domain containing 14; PR domain zinc finger protein 14; PR domain-containing 14; PR domain-containing protein 14
Gene Aliases: PFM11; PRDM14
UniProt ID: (Human) Q9GZV8
Entrez Gene ID: (Human) 63978
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support