Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KHCKRHTGI KDFMCELCGK TFSERNTMET HKLIHTVGKQ WTCSVCDKKY VTEYMLQKHV QLTHDKVEAQ SCQLCGTKVS TRASMSRHMR RKHPEVLAVR IDDLDHLPET TTIDASSIGI VQPELTLEQE DLAEGKHGKA AKRSHKRKQK PEEEAGAPVP EDATFSEYSE KETEFTGSVG DETNSAVQSI QQVVVTLGDP NVTTPSSSVG LTNITVTPIT TAAATQFTNL QPVAVGHLTT PERQLQLDNS ILTVTFDTVS GSAMLHNRQN DVQIHPQPEA SNPQSVAHFI NLTTLVNSIT PLGSQLSDQH PLTWRAVPQT DVLPPPQPQA PPQQAAQPQV QAEQQQQQMY SY (791-1141 aa encoded by BC067102)
The exact function of PRDM15 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PFM12; PR domain containing 15; PR domain zinc finger protein 15; PR domain-containing protein 15; Zinc finger protein 298
Gene Aliases: C21orf83; PFM15; PRDM15; ZNF298
UniProt ID: (Human) P57071
Entrez Gene ID: (Human) 63977
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support