Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive test controls include: Mouse spleen, Mouse stomach, Rat testis, Rat liver.
Immunogen sequence: DSEDSDEEWT PKQQVSPPWV PFRVKHSKQQ KESSRMPFSG ESNVKEGSGI ENLLNTSGSE HVQKPVSSLE EGNTSGQHSG KKLKLRKKNV EVKMYRLRER KGLAYEEVSE PQDDDYLYCE KCQNFFIDSC PNHGPPLFVK DSMVDRGHPN HSVLSLPPGL RISPSGIPEA GLGVWNEASD LPVGLHFGPY EGQITEDEEA ANSGYSWLIT KGRNCYEYVD GQDESQANWM RYVNCARDDE EQNLVAFQYH RKIFYRTCRV IRPGCELLVW YGDEYGQELG IKWG
The PR domain is a protein-protein interaction module of about 100 amino acids. PR domain-containing proteins, such as PRDM9, are often involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-lysine36 N-trimethyltransferase PRDM9; [histone H3]-lysine4 N-trimethyltransferase PRDM9; [histone H3]-lysine9 N-trimethyltransferase PRDM9; [histone H4]-lysine20 N-methyltransferase PRDM9; [histone H4]-N-methyl-L-lysine20 N-methyltransferase PRDM9; Histone-lysine N-methyltransferase PRDM9; hybrid sterility protein 1; meiosis-induced factor containing a PR/SET domain and zinc-finger motif; PR domain containing 7; PR domain zinc finger protein 9; PR domain-containing protein 9; PR-domain zinc finger protein; Protein-lysine N-methyltransferase PRDM9; VPR domain containing 9
Gene Aliases: BC012016; Dsbc1; G1-419-29; Meisetz; Prdm7; Prdm9; Rcr1; repro7
UniProt ID: (Rat) P0C6Y7
Entrez Gene ID: (Mouse) 213389, (Rat) 365155
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support