Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The PR domain is a protein-protein interaction module of about 100 amino acids. PR domain-containing proteins, such as PRDM9, are often involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-lysine36 N-trimethyltransferase PRDM9; [histone H3]-lysine4 N-trimethyltransferase PRDM9; [histone H3]-lysine9 N-trimethyltransferase PRDM9; [histone H4]-lysine20 N-methyltransferase PRDM9; [histone H4]-N-methyl-L-lysine20 N-methyltransferase PRDM9; Histone-lysine N-methyltransferase PRDM9; hybrid sterility protein 1; meiosis-induced factor containing a PR/SET domain and zinc-finger motif; PR domain containing 7; PR domain zinc finger protein 9; PR domain-containing protein 9; PR-domain zinc finger protein; Protein-lysine N-methyltransferase PRDM9; VPR domain containing 9
Gene Aliases: BC012016; Dsbc1; G1-419-29; Meisetz; Prdm7; Prdm9; Rcr1; repro7
UniProt ID: (Rat) P0C6Y7
Entrez Gene ID: (Mouse) 213389, (Rat) 365155
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support