Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACR1; Alu co-repressor 1; B166; Natural killer cell-enhancing factor A; natural killer-enhancing factor A; NKEF-A; PAG; Peroxiredoxin-1; proliferation-associated gene A; Proliferation-associated gene protein; Prx 1; Thioredoxin peroxidase 2; Thioredoxin-dependent peroxide reductase 2; Thioredoxin-dependent peroxiredoxin 1
Gene Aliases: MSP23; NKEF-A; NKEFA; PAG; PAGA; PAGB; PRDX1; PRX1; PRXI; TDPX2
UniProt ID: (Human) Q06830
Entrez Gene ID: (Human) 5052
Molecular Function:
peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support