Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSGNAKIG HPAPNFKATA VMPDGQFKDI SLSDYKGKYV VFFFYPLDFT FVCPTEIIAF SDRAEEFKKL NCQVIGASVD SHFCHLAWVN TPKKQGGLGP MNIPLVSDPK RTIAQDYGVL KADEGISFRG LFIIDDKGIL RQITVNDLPV GRSVDETLRL VQAFQFTDKH GEVCPAGWKP GSDTIKPDVQ KSKEYFSKQK (1-199 aa encoded by BC007063)
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACR1; Alu co-repressor 1; B166; HBP23; Heme-binding 23 kDa protein; macrophage 23 Kd stress protein; Macrophage 23 kDa stress protein; macrophage 23kDa stress protein; macrophase stress protein 22kDa; macrophase stress protein 23 kd; Natural killer cell-enhancing factor A; natural killer-enhancing factor A; NKEF-A; OSF-3; osteoblast specific factor 3; Osteoblast-specific factor 3; PAG; Peroxiredoxin-1; proliferation-associated gene A; Proliferation-associated gene protein; Prx 1; thioredoxin dependent peroxide reductase 2; Thioredoxin peroxidase 2; Thioredoxin-dependent peroxide reductase 2; Thioredoxin-dependent peroxiredoxin 1; Trx dependent peroxide reductase 2
Gene Aliases: Hbp23; MSP23; NKEF-A; NKEFA; OSF-3; OSF3; PAG; PAGA; PAGB; PRDX1; PrdxI; PRX1; PRXI; TDPX2; TDX2; TPxA
UniProt ID: (Human) Q06830, (Mouse) Q3UBV4, (Rat) Q63716
Entrez Gene ID: (Human) 5052, (Mouse) 18477, (Rat) 117254
Molecular Function:
peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support