Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: PFPLYALQV DPSTGLLIAA GGGGAAKTGI KNGVHFLQLE LINGRLSASL LHSHDTETRA TMNLALAGDI LAAGQDAHCQ LLRFQAHQQQ GNKAEKAGSK EQGPRQRKGA APAEKKCGAE TQHEGLELRV ENLQAVQTDF SSDPLQKVVC FNHDNTLLAT GGTDGYVRVW KVPSLEKVLE FKAHEGEIED L (13-202 aa encoded by BC002765)
Prolactin (PRL) expression in the pituitary is limited to specific cells. Pit-1 is a pituitary specific transcription factor that plays an important role in PRL expression, both in mature organism and during development. The PRL promoter contains numerous Pit-1 binding sites and these sites have been implicated in both basal level and kinase-mediated gene expression. The most proximal of these binding sites, termed 1P, has been shown to direct a response to numerous signals, such as cAMP and various G-proteins. A novel protein, termed PREB (prolactin regulatory element binding protein), has been recently identified that regulates PRL gene expression through the 1P site, though it contains no discernable DNA-binding motif. Recent studies suggest that PREB is encoded by a single-copy gene in both mice and humans and exhibits nuclear accumulation in pituitary cells. Evidence suggests that PREB is a novel transcription factor that assists in PRL expression whether alone, or in concert with Pit-1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Guanine nucleotide-exchange factor SEC12; mammalian guanine nucleotide exchange factor mSec12; Mammalian guanine nucleotide exchange factor Sec12; mSec12; prolactin regulatory binding-element protein; Prolactin regulatory element-binding protein
Gene Aliases: C85705; PREB; SEC12
UniProt ID: (Human) Q9HCU5, (Mouse) Q9WUQ2
Entrez Gene ID: (Human) 10113, (Mouse) 50907
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support