Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVKYFLGQS VLRSSWDQVF AAFWQRYPNP YSKHVLTEDI VHREVTPDQK LLSRRLLTKT NRMPRWAERL FPANVAHSVY VLEDSIVDPQ NQTMTTFTWN INHARLMVVE ERCVYCVNSD NSGWTEIRRE AWVSSSLFGV SRAVQEFGLA RFKSNVTKTM KGFEYILAKL QGEAPSKTLV ETAKEAKEKA KETALAATEK AKDLASKAAT KKQQQQQQFV (1-219 aa encoded by BC008307)
May be important for the development of vital and immunocompetent organs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 25 kDa protein of relevant evolutionary and lymphoid interest; PRELI domain-containing protein 1, mitochondrial; protein of relevant evolutionary and lymphoid interest; PX19 homolog; Px19-like protein
Gene Aliases: 2610524G07Rik; CGI-106; PRELI; PRELID1; PX19; RGD1308082; SBBI12
UniProt ID: (Human) Q9Y255, (Mouse) Q8R107
Entrez Gene ID: (Human) 27166, (Rat) 290995, (Mouse) 66494
Molecular Function:
immunoglobulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support