Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
May be important for the development of vital and immunocompetent organs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 25 kDa protein of relevant evolutionary and lymphoid interest; PRELI domain-containing protein 1, mitochondrial; protein of relevant evolutionary and lymphoid interest; PX19 homolog; Px19-like protein
Gene Aliases: 2610524G07Rik; CGI-106; PRELI; PRELID1; PX19; RGD1308082; SBBI12
UniProt ID: (Human) Q9Y255, (Mouse) Q8R107
Entrez Gene ID: (Human) 27166, (Rat) 290995, (Mouse) 66494
Molecular Function: immunoglobulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support