Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
PRELID2 (PRELI Domain Containing 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include phosphatidic acid transporter activity. An important paralog of this gene is PRELID3B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTC-806A22.1; PRELI domain-containing protein 2
Gene Aliases: PRELID2
UniProt ID: (Human) Q8N945
Entrez Gene ID: (Human) 153768
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support