Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLSLQYPDVY RDETAVQDYH GHKICDPYAW LEDPDSEQTK AFVEAQNKIT VPFLEQCPIR GLYKERMTEL YDYPKYSCHF KKGKRYFYFY NTGLQNQRVL YVQDSLEGEA RVFLDPNILS DDGTVALRGY AFSEDGEYFA YGLSASGSDW VTIKFMKVDG AKELPDVLER VKFSCMAWTH DGKGMFYNSY PQQDGKSDGT ETSTNLHQKL YYHVLGTDQS EDILCAEFPD EPKWMGGAEL SDDGRYVLLS IREGCDPVNR LWYCDLQQES SGIAGILKWV KLIDNFEGEY DYVTNEGTVF TFKTNRQSPN YRVINIDFRD PEESKWKVLV PEHEKDVLEW IACVRSNFLV LCYLHDVKNI LQLHDLTTGA LLKTFPLDVG SIVGYSGQKK DTEIFYQFTS FLSPGIIYHC DLTKEELEPR VFREVTVKGI DASDYQTVQI FYPSKDGTKI PMFIVHKKGI KLDGSHPAFL YGYGGFNISI TPNYSVSRLI FVRHMGGILA VANIRGGGEY GETWHKGGIL ANKQNCFDDF QCAAEYLIKE GYTSPKRLTI NGGSNGGLLV AACANQRPDL FGCVIAQVGV MDMLKFHKYT IGHAWTTDYG CSDSKQHFEW LVKYSPLHNV KLPEADDIQY PSMLLLTADH DDRVVPLHSL KFIATLQYIV GRSRKQSNPL LIHVDTKAGH GAGKPTAKVI EEVSDMFAFI ARCLNVDWIP
p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as does cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ355L5.1 (prolyl endopeptidase); EC 3.4.21.26; MGC16060; PE; Post-proline cleaving enzyme; Prolyl endopeptidase; prolyl oligopeptidase
Gene Aliases: PE; PEP; PREP
UniProt ID: (Human) P48147
Entrez Gene ID: (Human) 5550
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support