Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDLKMNFRP ERRVLVDDGW ILAYCHVRYG GELGLQWHAD GRLTKKLNGL ADLEACIKTL HGQGFSQPSL TTLTAFSAGG VLAGALCNSN PELVRAVTLE APFLDVLNTM MDTTLPLTLE ELEEWGNPSS DEKHKNYIKR YCPYQNIKPQ HYPSIHITAY ENDERVPLKG IVSYTEKLKE AIAEHAKDTG EGYQTPNIIL DIQPGGNHVI EDSHKKITAQ IKFLYEELGL DSTSVFEDLK KYLKF (1-244 aa encoded by BC013193)
The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Prolyl endopeptidase-like; Prolylendopeptidase-like; putative prolyl oligopeptidase
Gene Aliases: 2810457N15Rik; 9530014L06Rik; D030028O16Rik; KIAA0436; mKIAA0436; PREPL; RGD1310143
UniProt ID: (Human) Q4J6C6, (Mouse) Q8C167, (Rat) Q5HZA6
Entrez Gene ID: (Human) 9581, (Mouse) 213760, (Rat) 298771
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support