Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LIFCSRACSLG SEPTAPGPSR RSWSAGPVTA PLAASTASFS AVKGASETTT KGTSTELAPA TGPEEPSRFL RGAPHRHSMP ELGLRSVPEP PPESPGQPNL RPDDSAFGRQ S
Highest antigen sequence identity to the following orthologs - mouse 79%, rat 75%.
LIM domain only 6 is a three LIM domain-containing protein. The LIM domain is a cysteine-rich sequence motif that binds zinc atoms to form a specific protein-binding interface for protein-protein interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LIM domain only protein 6; LMO-6; Pk3; prickle homolog 3; Prickle planar cell polarity protein 3; Prickle-like protein 3; Triple LIM domain protein 6
Gene Aliases: LMO6; PRICKLE3
UniProt ID: (Human) O43900
Entrez Gene ID: (Human) 4007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support