Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MAPFLRIAFN SYELGSLQAE DEANQPFCAV KMKEALSTER GKTLVQKKPT MYPEWKSTFD AHIYEGRVIQ IVLMRAAEEP VSEVTVGVSV LAERCKKNNG KAEFWLDLQP QAKVLMSVQY FLEDVDCKQS MRSEDEAKFP TMNRRGAIKQ AKIHYIKNHE FIATFFGQPT FCSVCKDFVW GLNKQGYKCR QCNAAIHKKC IDKIIGRCTG TAANSRDTIF QKERFNIDMP HRFKVHNYMS PTFCDHCGSL LWGLVKQGLK CEDCGMNVHH KCREKVANLC GINQKLLAEA LNQVTQRASR RSDSASSEPV GIYQGFEKKT GVAGEDMQDN SGTYGKIWEG SSKCNINNFI FHKVLGKGSF GKVLLGELKG RGEYFAIKAL KKDVVLIDDD VECTMVEKRV LTLAAENPFL THLICTFQTK DHLFFVMEFL NGGDLMYHIQ DKGRFELYRA TFYAAEIMCG LQFLHSKGII YRDLKLDNVL LDRDGHIKIA DFGMCKENIF GESRASTFCG TPDYIAPEIL QGLKYTFSVD WWSFGVLLYE MLIGQSPFHG DDEDELFESI RVDTPHYPRW ITKESKDILE KLFEREPTKR LGVTGNIKIH PFFKTINWTL LEKRRLEPPF RPKVKSPRDY SNFDQEFLNE KARLSYSDKN LIDSMDQSAF AGFSFVNPKF EHLLED
The PKC family of serine/threonine kinases, including PRKCD (PKC delta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II, and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. Protein kinase C (PKC) can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. The protein encoded by this gene is one of the PKC family members. Studies both in human and mice demonstrate that this kinase is involved in B cell signaling and in the regulation of growth, apoptosis, and differentiation of a variety of cell types. Alternatively spliced transcript variants encoding the same protein have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-May; kinase nPKC-delta; KPCD; nPKC-delta; Protein kinase C delta type; protein kinase C delta VIII; protein kinase C, delta; Tyrosine-protein kinase PRKCD
Gene Aliases: ALPS3; CVID9; MAY1; nPKC-delta; PKCD; PRKCD
UniProt ID: (Human) Q05655
Entrez Gene ID: (Human) 5580
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support