Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the "Signalosome", which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Caveolae-associated protein 3; cavin 3; Cavin-3; D3T-inducible gene 2 protein; DIG-2; Dithiolethione-inducible gene 2 protein; dithiolethione-inducible gene-2; hSRBC; MGC20400; PKC-delta binding protein; Protein kinase C delta-binding protein; protein kinase C, delta binding protein; sdr-elated gene product that binds to c-kinase; sdr-related gene product that binds to c-kinase; Serum deprivation response factor-related gene product that binds to C-kinase
Gene Aliases: 3110015B12Rik; 6330514M23Rik; cavin-3; CAVIN3; DIG-2; Dig2; HSRBC; PRKCDBP; SRBC
UniProt ID: (Human) Q969G5, (Mouse) Q91VJ2, (Rat) Q9Z1H9
Entrez Gene ID: (Human) 112464, (Mouse) 109042, (Rat) 85332
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support