Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MSHTVAGGGS GDHSHQVRVK AYYRGDIMIT HFEPSISFEG LCNEVRDMCS FDNEQLFTMK WIDEEGDPCT VSSQLELEEA FRLYELNKDS ELLIHVFPCV PERPGMPCPG EDKSIYRRGA RRWRKLYCAN GHTFQAKRFN RRAHCAICTD RIWGLGRQGY KCINCKLLVH KKCHKLVTIE CGRHSLPQEP VMPMDQSSMH SDHAQTVIPY NPSSHESLDQ VGEEKEAMNT RESGKASSSL GLQDFDLLRV IGRGSYAKVL LVRLKKTDRI YAMKVVKKEL VNDDEDIDWV QTEKHVFEQA SNHPFLVGLH SCFQTESRLF FVIEYVNGGD LMFHMQRQRK LPEEHARFYS AEISLALNYL HERGIIYRDL KLDNVLLDSE GHIKLTDYGM CKEGLRPGDT TSTFCGTPNY IAPEILRGED YGFSVDWWAL GVLMFEMMAG RSPFDIVGSS DNPDQNTEDY LFQVILEKQI RIPRSMSVKA ASVLKSFLNK DPKERLGCLP QTGFADIQGH PFFRNVDWDM MEQKQVVPPF KPNISGEFGL DNFDSQFTNE RVQLTPDDDD IVRKIDQSEF EGFEYINPLL MSAEECV
The PKC family of serine/threonine kinases, including PRKCI (PKC iota), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. The PKC family comprises at least eight members, which are differentially expressed and are involved in a wide variety of cellular processes. This protein kinase is calcium-independent and phospholipid-dependent. It is not activated by phorbolesters or diacylglycerol. This kinase can be recruited to vesicle tubular clusters (VTCs) by direct interaction with the small GTPase RAB2, where this kinase phosphorylates glyceraldehyde-3-phosphate dehydrogenase (GAPD/GAPDH) and plays a role in microtubule dynamics in the early secretory pathway. This kinase is found to be necessary for BCL-ABL-mediated resistance to drug-induced apoptosis and therefore protects leukemia cells against drug-induced apoptosis. There is a single exon pseudogene mapped on chromosome X.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aPKC-lambda/iota; Atypical protein kinase C-lambda/iota; nPKC-iota; PRKC-lambda/iota; Protein kinase C iota type; protein kinase C, iota
Gene Aliases: DXS1179E; nPKC-iota; PKCI; PRKCI
UniProt ID: (Human) P41743
Entrez Gene ID: (Human) 5584
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support