Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSPFLRIGLS NFDCGSCQSC QGEAVNPYCA VLVKEYVESE NGQMYIQKKP TMYPPWDSTF DAHINKGRVM QIIVKGKNVD LISETTVELY SLAERCRKNN GKTEIWLELK PQGRMLMNAR YFLEMSDTKD MNEFETEGFF ALHQRRGAIK QAKVHHVKCH EFTATFFPQP TFCSVCHEFV WGLNKQGYQC RQCNAAIHKK CIDKVIAKCT GSAINSRETM FHKERFKIDM PHRFKVYNYK SPTFCEHCGT LLWGLARQGL KCDACGMNVH HRCQTKVANL CGINQKLMAE ALAMIESTQQ ARCLRDTEQI FREGPVEIGL PCSIKNEARP PCLPTPGKRE PQGISWESPL DEVDKMCHLP EPELNKERPS LQIKLKIEDF ILHKMLGKGS FGKVFLAEFK KTNQFFAIKA LKKDVVLMDD DVECTMVEKR VLSLAWEHPF LTHMFCTFQT KENLFFVMEY LNGGDLMYHI QSCHKFDLSR ATFYAAEIIL GLQFLHSKGI VYRDLKLDNI LLDKDGHIKI ADFGMCKENM LGDAKTNTFC GTPDYIAPEI LLGQKYNHSV DWWSFGVLLY EMLIGQSPFH GQDEEELFHS IRMDNPFYPR WLEKEAKDLL VKLFVREPEK RLGVRGDIRQ HPLFREINWE ELERKEIDPP FRPKVKSPFD CSNFDKEFLN EKPRLSFADR ALINSMDQNM FRNFSFMNPG MERLIS
Protein Kinase C (PKC) isoforms are serine/threonine kinases involved in signal transduction pathways that govern a wide range of physiological processes including differentiation, proliferation, gene expression, brain function, membrane transport and the organization of cytoskeletal and extracellular matrix proteins. Increasing evidence from studies using in vitro and in vivo systems points to PKC as a key regulator of critical cell cycle transitions, including cell cycle entry and exit and the G1 and G2 checkpoints. PKC-mediated control of these transitions can be negative or positive, depending on the timing of PKC activation during the cell cycle and on the specific PKC isozymes involve. There have been at least 12 different PKC isoforms identified in humans to date, including alpha, beta I, beta II, gamma, delta, epsilon, zeta, eta, theta, iota, lambda, and mu. The different PKC isoforms vary in structure, subcellular localization, tissue distribution and response to extracellular signals. PKC beta II has been shown to be expressed, in part, in cardiac muscle, intestinal epithelium, kidney and pancreas. PKC beta II has been shown to be involved in cell cycle progression and cancer proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.7.11.13; kinase nPKC-theta; KPCT; MGC126514; MGC141919; nPKC-theta; Protein kinase C theta type; protein kinase C, theta; RP11-563J2.1
Gene Aliases: nPKC-theta; PRKCQ; PRKCT
UniProt ID: (Human) Q9H549
Entrez Gene ID: (Human) 5588
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support