Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MSANNSPPS AQKSVLPTAI PAVLPAASPC SSPKTGLSAR LSNGSFSAPS LTNSRGSVHT VSFLLQIGLT RESVTIEAQE LSLSAVKDLV CSIVYQKFPE CGFFGMYDKI LLFRHDMNSE NILQLITSTD EIHEGDLVEV VLSALATVED FQIRPHTLYV HSYKAPTFCD YCGEMLWGLV RQGLKCEGCG LNYHKRCAFK IPNNCSGVRK RRLSNVSLPG PGLSVPRPLQ PEYVALPSEE SHVHQEPSKR IPSWSGRPIW MEKMVMCRVK VPHTFAVHSY TRPTICQYCK RLLKGLFRQG MQCKDCKFNC HKRCASKVPR DCLGEVTFNG EPSSLGTDTD IPMDID (1-345 aa encoded by B C030706)
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role. The protein encoded by this gene is one of the PKC family members. This kinase can be activated rapidly by the agonists of G protein-coupled receptors. It resides in both cytoplasm and nucleus, and its nuclear accumulation is found to be dramatically enhanced in response to its activation. This kinase can also be activated after B-cell antigen receptor (BCR) engagement, which requires intact phopholipase C gamma and the involvement of other PKC family members.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: nPKC-nu; Protein kinase C nu type; protein kinase C, nu; Protein kinase EPK2; Serine/threonine-protein kinase D3
Gene Aliases: 4930557O20Rik; 5730497N19Rik; EPK2; nPKC-NU; PKC-NU; Pkcnu; PKD3; PRKCN; PRKD3
UniProt ID: (Human) O94806, (Mouse) Q8K1Y2
Entrez Gene ID: (Human) 23683, (Rat) 313834, (Mouse) 75292
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support