Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody is specific to PRKG2.
PRKG2 (PKG2, cGKII) exhibits many cellular functions and has cAMP- and cGMP-dependent protein kinase activity. cGKII is thought to play a key role in a diverse set of physiological pathway. cGKII may mediate intestinal secretion of water and electrolytes induced by the E. coli toxin STa and the intestinal peptide guanylin. Identification of the pathway that mediates intestinal fluid secretion by E. coli. cGKII is postulated to be a molecular switch that couples the cessation of proliferation and the start of hypertrophic chondrocyte differentiation through attenuating SOX9 function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CGK 2; cGK2; cGKII; cGMP dependent protein kinase type II; cGMP-dependent protein kinase 2; cGMP-dependent protein kinase II; EC 2.7.11.12; protein kinase cGMP- dependent type 2; protein kinase, cGMP- dependent, type 2; protein kinase, cGMP- dependent, type II; testicular tissue protein Li 99; type II cGMP-dependent protein kinase
Gene Aliases: AW212535; cGK-II; cGK2; CGKII; GDPKII; PRKG2; PRKGR2
UniProt ID: (Human) Q13237, (Rat) Q64595
Entrez Gene ID: (Human) 5593, (Rat) 25523, (Mouse) 19092
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support