Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • PACT Antibodies

            Abnova

            PRKRA Monoclonal Antibody (1B9-1A7)

            View all (16) PACT antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # H00008575-M01
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            455.00
            100 µg
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite PRKRA Monoclonal Antibody (1B9-1A7)

            • Antibody Testing Data (7)
            Application
            PRKRA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 7

            PRKRA Antibody (H00008575-M01) in IHC (P)

            Immunoperoxidase of monoclonal antibody to PRKRA on formalin-fixed paraffin-embedded human squamous cell carcinoma. Antibody concentration is 5 µg/mL.
            View Product

            PRKRA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            PRKRA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            PRKRA Antibody in Western Blot (WB)
            PRKRA Antibody in Western Blot (WB)
            PRKRA Antibody in Western Blot (WB)
            PRKRA Antibody in Immunoprecipitation (IP)
            PRKRA Antibody in ELISA (ELISA)

            Product Details

            H00008575-M01

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1-5 µg/mL
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            5 µg/mL
            -

            ELISA (ELISA)

            0.03 ng/mL
            -

            Immunoprecipitation (IP)

            1-5 µg/mL
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG1, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            1B9-1A7

            Immunogen

            PRKRA (AAH09470, 1 a.a. approximately 313 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            See Label

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.4

            Contains

            no preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            Sequence of this protein is as follows: MSQSRHRAEA PPLEREDSGT FSLGKMITAK PGKTPIQVLH EYGMKTKNIP VYECERSDVQ IHVPTFTFRV TVGDITCTGE GTSKKLAKHR AAEAAINILK ANASICFAVP DPLMPDPSKQ PKNQLNPIGS LQELAIHHGW RLPEYTLSQE GGPAHKREYT TICRLESFME TGKGASKKQA KRNAAEKFLA KFSNISPENH ISLTNVVGHS LGCTWHSLRN SPGEKINLLK RSLLSIPNTD YIQLLSEIAK EQGFNITYLD IDELSANGQY QCLAELSTSP ITVCHGSGIS CGNAQSDAAH NALQYLKIIA ERK

            Target Information

            PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: interferon-inducible double stranded RNA-dependent protein kinase activator A; Interferon-inducible double-stranded RNA-dependent protein kinase activator A; PKR-associated protein X; PKR-associating protein X; Protein activator of the interferon-induced protein kinase; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; Protein kinase, interferon-inducible double-stranded RNA-dependent activator

            View more View less

            Gene Aliases: DYT16; HSD-14; HSD14; PACT; PRKRA; RAX

            View more View less

            UniProt ID: (Human) O75569

            View more View less

            Entrez Gene ID: (Human) 8575

            View more View less

            Function(s)

            double-stranded RNA binding protein binding enzyme activator activity enzyme binding identical protein binding protein homodimerization activity poly(A) RNA binding

            Process(es)

            protein phosphorylation immune response negative regulation of cell proliferation response to virus production of siRNA involved in RNA interference gene silencing by RNA pre-miRNA processing cellular response to oxidative stress production of miRNAs involved in gene silencing by miRNA outer ear morphogenesis middle ear morphogenesis positive regulation of catalytic activity skeletal system morphogenesis positive regulation of intrinsic apoptotic signaling pathway
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-njtcd:80/100.66.77.138:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0